missing translation for 'onlineSavingsMsg'
Learn More

Poulten & Graf GmbH VOLAC FORTUNA™ Graduated Pipette with Blue Graduation

Product Code. 15972100
Click to view available options
Volume (Metric):
1 mL
2 mL
5 mL
10 mL
20 mL
Unit Size:
Each
Pack of 5
This item is not returnable. View return policy

Product Code. 15972100

Brand: Poulten & Graf GmbH 1 11441402

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Poulten & Graf GmbH VOLAC FORTUNA™ Graduated Pipette with Blue Graduation is ideal for measuring and transferring various quantities of liquid.

  • Total delivery
  • Blue graduation
  • With ring marks
  • Comformity approved

Specifications

Certifications/Compliance EN ISO 835
Class AS
Description Graduated pipette
Material Soda Lime Glass
Sterility Non-sterile
Graduated Yes
Graduation Color Blue
Tolerance 0.10 mL
Volume (Metric) 20 mL
Type Graduated Pipet
Disposable Reusable
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.