Learn More
Poulten & Graf GmbH VOLAC FORTUNA™ Graduated Pipette with Single Certificate
Poulten & Graf GmbH VOLAC FORTUNA™ Graduated Pipette with Single Certificate is ideal for measuring and transferring various quantities of liquid.
€ 18.35 - € 35.60
Specifications
Material | Soda Lime Glass |
---|---|
Certifications/Compliance | EN ISO 835 |
Class | AS |
Disposable | No |
Autoclavable | Yes |
Product Code | Brand | Capacity (Metric) | Graduations | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Graduations | Price | Quantity & Availability | ||||
15559411
|
Poulten & Graf GmbH
R385/WAC/A/5 |
1 mL | 0.01 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15569411
|
Poulten & Graf GmbH
R385/WAC/B/5 |
2 mL | 0.02 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15579411
|
Poulten & Graf GmbH
R385/WAC/C/5 |
5 mL | 0.05 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15589411
|
Poulten & Graf GmbH
R385/WAC/D/5 |
10 mL | 0.10 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15599411
|
Poulten & Graf GmbH
R385/WAC/E/5 |
25 mL | 0.10 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Total delivery
- Conformity approved
- Zero at top
- Amber graduation
- With ring marks
- Non blowout
- With single certificate
Specifications
Soda Lime Glass | |
AS | |
Yes |
EN ISO 835 | |
No |