Learn More
Poulten & Graf GmbH VOLAC USP Graduated Cylinder
Safely and accurately aquire measurements for all laboratory needs. Poulten & Graf GmbH VOLAC USP Graduated Cylinder measures the volume of a liquid using horizontal marked lines.
€ 165.00 - € 500.00
Specifications
Base Type | Hexagonal |
---|---|
Calibration | To Contain |
Certifications/Compliance | EN ISO 4788 |
Class | A |
Type | Graduated Cylinder |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10739804
|
Poulten & Graf GmbH
US263/WAC/D/2 |
25 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10224002
|
Poulten & Graf GmbH
US263/WAC/A/2 |
5 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10496562
|
Poulten & Graf GmbH
US263/WAC/F/2 |
100 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10623673
|
Poulten & Graf GmbH
US263/WAC/I/2 |
1000 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10486562
|
Poulten & Graf GmbH
US263/WAC/B/2 |
10 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10613673
|
Poulten & Graf GmbH
US263/WAC/H/2 |
500 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10395592
|
Poulten & Graf GmbH
US263/WAC/J/S |
2000 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10406572
|
Poulten & Graf GmbH
US263/WAC/G/2 |
150 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Hexagonal base
- White graduation
- With individual serial number
- With single certificate
Specifications
Hexagonal | |
EN ISO 4788 | |
Graduated Cylinder | |
Round | |
White |
To Contain | |
A | |
Blue | |
Borosilicate Glass | |
Autoclavable |