Learn More
Poulten & Graf GmbH VOLAC USP Mixing Cylinder
Click to view available options
Capacity (Metric):
10 mL
50 mL
250 mL
500 mL
1000 mL
2000 mL
Graduations:
0.2 mL
1 mL
2 mL
5 mL
10 mL
Unit Size:
Each
Pack of 2
Description
- With PP-stopper
- Hexagonal base
- White graduation
- With individual serial number
- With single certificate
Specifications
Specifications
| Item Description | Volac Mixing Cylinder, USP, with single certificate |
| Base Type | Hexagonal |
| Calibration | To Contain |
| Scale Color | Blue |
| Shape | Round |
| Type | Graduated Mixing Cylinder |
| Tolerance | ±0.08 mL |
| Material | Borosilicate Glass |
| Certifications/Compliance | EN ISO 4788 |
| Class | A |
| Show More |