Learn More
Invitrogen™ Bemarituzumab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA558313
This item is not returnable.
View return policy
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Bemarituzumab is a subtype-selective humanized monoclonal antibody against FGFR2b. The FIGH study aims to evaluate the efficacy of its combined chemotherapy for untreated advanced gastric cancer or gastroesophageal junction cancer.
Specifications
| Bemarituzumab | |
| Recombinant Monoclonal | |
| Unconjugated | |
| FPA114-A; FPA-144 | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| 1 mg/mL | |
| 25mM Histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Human | |
| 100 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |