missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Volociximab Recombinant Monoclonal Antibody

Recombinant Monoclonal Antibody
€ 477.00 - € 1204.00
Specifications
| Antigen | Volociximab Chimeric |
|---|---|
| Concentration | 1 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30282754
![]() |
Invitrogen™
MA559893 |
100 μg |
€ 477.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30284599
|
Invitrogen™
MA559894 |
1 mg |
€ 1204.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Volociximab is a biosimilar that targets ITGA5.Specifications
| Volociximab Chimeric | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Lyophilized | |
| IgG4SP | |
| Human | |
| 558480-40-3; M20 | |
| Antibody |
| 1 mg/mL | |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| Unconjugated | |
| Human | |
| RUO | |
| 25 mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Primary | |
| Protein A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title