missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buerkle™ Volume Head for PharmaPicker
Volume tips to insert into the PharmaPicker collection cylinder.
Brand: Buerkle™ 5346-2130
Additional Details : Weight : 0.01000kg
Benachrichtigungen:
Further sampling volumes available on request!
Beschreibung
- Volume tips to insert into the PharmaPicker collection cylinder
- Sampling quantity 0.1 - 1.2 ml for collection cylinder 5346-1000
- Sampling quantity 1.25 - 2.5 ml for collection cylinder 5346-2000
- Stainless steel AISI 316L (1.4404)
Spezifikation
Volume Head | |
Stainless Steel (V4A) | |
1.3 mL | |
PharmaPicker Collection Cylinder |