Learn More
Buerkle™ Volume Head for PharmaPicker
Product Code. 11712493 Shop All Bürkle Products
Click to view available options
Capacity (Metric):
0.1 mL
0.2 mL
0.6 mL
1 mL
1.25 mL
2 mL
2.5 mL
Unit Size:
Each
Further sampling volumes available on request!
Product Code. 11712493
Brand: Buerkle™ 53462125
Only null left
Add to basket
Add to basket
Further sampling volumes available on request!
Volume tips to insert into the PharmaPicker collection cylinder.
  • Volume tips to insert into the PharmaPicker collection cylinder
  • Sampling quantity 0.1 to 1.2 mL for collection cylinder 5346-1000
  • Sampling quantity 1.25 to 2.5 mL for collection cylinder 5346-2000
  • Stainless steel AISI 316L (1.4404)
TRUSTED_SUSTAINABILITY
Specifications
Product Type Volume Head
Material Stainless Steel (V4A)
Autoclavable Autoclavable
Capacity (Metric) 1.25 mL
Finish Electro-polished
For Use With (Equipment) PharmaPicker