Learn More
Buerkle™ Volume Head for PharmaPicker
Shop All Bürkle Products
Click to view available options
Capacity (Metric):
0.1 mL
0.2 mL
0.6 mL
1 mL
1.25 mL
2 mL
2.5 mL
Unit Size:
Each
Description
- Volume tips to insert into the PharmaPicker collection cylinder
- Sampling quantity 0.1 to 1.2 mL for collection cylinder 5346-1000
- Sampling quantity 1.25 to 2.5 mL for collection cylinder 5346-2000
- Stainless steel AISI 316L (1.4404)
Specifications
Specifications
| Product Type | Volume Head |
| Material | Stainless Steel (V4A) |
| Capacity (Metric) | 2 mL |
| For Use With (Equipment) | PharmaPicker Collection Cylinder |