Learn More
Socorex™ Acura™ 8-channel Micropipette Volumetric Module
Product Code. 10315732
Click to view available options
Volume (Metric):
10 μL
50 μL
350 μL
Unit Size:
Each
Product Code. 10315732
missing translation for 'mfr': Socorex™ 800.08.010
Only null left
Add to basket
Add to basket
Increase the versatility of your Acura™ electronic micropipette control unit and save costs by accessorizing it with one of 27 available volumetric modules. Socorex™ 8-channel Micropipette Volumetric Module is an excellent and easily attachable/detachable choice for your multi-channel micropipetting needs.
  • One of 27 available volumetric assemblies.
    • Increase versatility and save costs.
  • Disassembles and reassembles without tools.
  • Calibration specificities of extra modules - as set by factory QC or by pipette owner - are retained in instrument memory.
Specifications
For Use With 8 Channel Pipette
Number of Channels 8
Product Line Acura
Type Micropipette Volumetric Module
Volume (Metric) 10 μL
For Use With (Application) 8 Channel Pipette
For Use With (Equipment) Acura™ Electro 926 Micropipette
Quantity 1 Each