Learn More
Witeg™ Volumetric Pipets, Blow-Out, Class B
Enjoy the durability of tempered tips that offer a wear resistance up to seven times better than conventional pipets. Witeg™ Volumetric Pipets, Blow-Out, Class B have amber DIFFICO ring marks and inscriptions.
Brand: Witeg™ 3 571 050
Additional Details : Weight : 0.02400kg
Description
- Blow-out type.
- Class B without waiting time.
- Tempered tips.
Specifications
DIN EN ISO 648 | |
Yes | |
5 mL | |
12 Pieces | |
5 mL | |
White | |
Volumetric Pipet |
B | |
Amber | |
Without waiting time | |
Blowout | |
Pack | |
5 mL |