missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Von Hippel Lindau Antibody (OTI1E1), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74851UV
This item is not returnable.
View return policy
Description
Von Hippel Lindau Monoclonal specifically detects Von Hippel Lindau in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Von Hippel Lindau | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| VHL | |
| Full length recombinant protein of human Von Hippel Lindau (NP_000542) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2b |
| Western Blot, Immunohistochemistry | |
| OTI1E1 | |
| Western Blot, Immunohistochemistry | |
| elongin binding protein, HRCA1, Protein G7, pVHL, RCA1, VHL1, von Hippel-Lindau disease tumor suppressor, von Hippel-Lindau syndrome, von Hippel-Lindau tumor suppressor | |
| Mouse | |
| Affinity Purified | |
| Apoptosis, Cancer, HIF Target Genes, Hypoxia, Neuronal Cell Markers, Neuronal Stem Cell Markers | |
| 7428.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction