missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Von Willebrand Factor Antibody (F8/44/20) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 201.00 - € 370.00
Specifications
| Antigen | Von Willebrand Factor |
|---|---|
| Clone | F8/44/20 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 |
| Applications | Western Blot, ELISA, Immunohistochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18635958
|
Novus Biologicals
NBP2-50362-0.025ml |
0.025 mL |
€ 201.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18635768
|
Novus Biologicals
NBP2-50362 |
0.1 mL |
€ 370.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Von Willebrand Factor Monoclonal antibody specifically detects Von Willebrand Factor in Human samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
| Von Willebrand Factor | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Cancer | |
| PBS | |
| 7450 | |
| IgG1 | |
| Protein A purified |
| F8/44/20 | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| coagulation factor VIII VWF, F8, F8VWF, von Willebrand factor, VWD, vWF | |
| Von Willebrand factor isolated from human plasma | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title