missing translation for 'onlineSavingsMsg'
Learn More

Von Willebrand Factor Antibody (OTI6C7), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-74852FR

Product Code. 18184747

  • € 751.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Von Willebrand Factor Monoclonal specifically detects Von Willebrand Factor in Human samples. It is validated for Western Blot, Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Von Willebrand Factor
Monoclonal
DyLight 680
PBS with 0.05% Sodium Azide
VWF
Recombinant protein fragment corresponding to amino acids 764-1263 of human Von Willebrand Factor (NP_000543) produced in SF9 cell.
0.1 mL
Primary
Human
IgG1
Western Blot, Immunohistochemistry
OTI6C7
Western Blot, Immunohistochemistry
coagulation factor VIII VWF, F8, F8VWF, von Willebrand factor, VWD, vWF
Mouse
Affinity Purified
Cancer
7450.0
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.