missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Von Willebrand Factor Antibody (VWF/1859R), PerCP, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-08963PCP
This item is not returnable.
View return policy
Description
Von Willebrand Factor Monoclonal antibody specifically detects Von Willebrand Factor in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Specifications
| Von Willebrand Factor | |
| Monoclonal | |
| PerCP | |
| coagulation factor VIII VWF, F8, F8VWF, von Willebrand factor, VWD, vWF | |
| Recombinant fragment of human Von Willebrand Factor protein (aa1815-1939) (exact sequence is proprietary) (Uniprot: P04275) | |
| 0.1 mL | |
| Cancer | |
| 7450 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry (Paraffin) | |
| VWF/1859R | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction