Learn More
Scientific Industries SI™ TurboMix Attachment
Click to view available options
Holds:
12 x 1.5 to 2 mL Tubes
Unit Size:
Each
Description
- Permanently mounts to most existing Vortex-Genie 2 family of mixers and the Vortex-Genie Pulse.
- Holds up to twelve 1.5mL or 2.0mL microtubes
- Ideal for aggressive and rapid dissolution, resuspension or mixing of samples
- Convenient and effective for cell disruption in glass bead procedures.
Specifications
Specifications
| Holds | 12 x 1.5 to 2 mL Tubes |
| Product Type | TurboMix Attachment |
| For Use With (Equipment) | Most Vortex-Genie 2 family of mixers, Vortex-Genie Pulse |
| No. of Positions | 12 |