missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VP1 Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-93575FR
This item is not returnable.
View return policy
Description
VP1 Polyclonal antibody specifically detects VP1 in Adeno-associated Virus (AAV) samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence
Specifications
| VP1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| DyLight 680 | |
| AAV, AAV2, Adeno-Associated Virus, Adeno-Associated Virus (AAV) | |
| Synthetic peptide within residues 70-90 of major coat protein VP3 [AAV 2], residues 270-290 of capsid protein VP1 [AAV 6], and residues 270-290 of capsid [AAV 3]. | |
| 0.1 mL | |
| Gene Therapy, Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites | |
| Adeno-associated Virus (AAV) | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction