missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VP26B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-12624-100ul
This item is not returnable.
View return policy
Description
VP26B Polyclonal antibody specifically detects VP26B in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| VP26B | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunoprecipitation 5-10uL/mg of lysate | |
| MGC10485, Pep8b, vacuolar protein sorting 26 homolog B (S. pombe), vacuolar protein sorting 26 homolog B (yeast), vacuolar protein sorting-associated protein 26B, Vesicle protein sorting 26B | |
| Produced in rabbits immunized with E. coli-derived Human VP26B fragment | |
| 100 μg | |
| Signal Transduction | |
| 112936 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| PBS, pH7.0 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction