missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VPRBP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599967
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total VPRBP.
Acts both as a substrate recognition component of E3 ubiquitin-protein ligase complexes and as an atypical serine/threonine-protein kinase, playing key roles in various processes such as cell cycle, telomerase regulation and histone modification. Probable substrate-specific adapter of a DCX (DDB1-CUL4-X-box) E3 ubiquitin-protein ligase complex, named CUL4A-RBX1-DDB1-DCAF1/VPRBP complex, which mediates ubiquitination and proteasome-dependent degradation of proteins such as NF2. Involved in the turnover of methylated proteins: recognizes and binds methylated proteins via its chromo domain, leading to ubiquitination of target proteins by the RBX1-DDB1-DCAF1/VPRBP complex. The CUL4A-RBX1-DDB1-DCAF1/VPRBP complex is also involved in B-cell development: VPRBP is recruited by RAG1 to ubiquitinate proteins, leading to limit error-prone repair during V(D)J recombination. Also part of the EDVP complex, an E3 ligase complex that mediates ubiquitination of proteins such as TERT, leading to TERT degradation and telomerase inhibition. Also acts as an atypical serine/threonine-protein kinase that specifically mediates phosphorylation of 'Thr-120' of histone H2A (H2AT120ph) in a nucleosomal context, thereby repressing transcription. H2AT120ph is present in the regulatory region of many tumor suppresor genes, down-regulates their transcription and is present at high level in a number of tumors. Involved in JNK-mediated apoptosis during cell competition process via its interaction with LLGL1 and LLGL2. In case of infection by HIV-1 virus, it is recruited by HIV-1 Vpr in order to hijack the CUL4A-RBX1-DDB1-DCAF1/VPRBP function leading to arrest the cell cycle in G2 phase, and also to protect the viral protein from proteasomal degradation by another E3 ubiquitin ligase. The HIV-1 Vpr protein hijacks the CUL4A-RBX1-DDB1-DCAF1/VPRBP complex to promote ubiquitination and degradation of proteins such as TERT and ZIP/ZGPAT. In case of infection by HIV-2 virus, it is recruited by HIV-2 Vpx in order to hijack the CUL4A-RBX1-DDB1-DCAF1/VPRBP function leading to enhanced efficiency of macrophage infection and promotion of the replication of cognate primate lentiviruses in cells of monocyte/macrophage lineage.
Specifications
| VPRBP | |
| Polyclonal | |
| Unconjugated | |
| DCAF1 | |
| AI447437; B930007L02Rik; DCAF1; DDB1 and CUL4 associated factor 1; DDB1- and CUL4-associated factor 1; HIV-1 Vpr binding protein; HIV-1 Vpr-binding protein; KIAA0800; LOW QUALITY PROTEIN: DDB1- and CUL4-associated factor 1; MGC102804; mKIAA0800; Protein VPRBP; protein VPRBP; DDB1- and CUL4-associated factor 1; RGD1305176; RIP; Serine/threonine-protein kinase VPRBP; Vpr (HIV-1) binding protein; Vpr-binding protein; VprBP; vpr-interacting protein | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 315987, 321006, 9730 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q80TR8, Q9Y4B6 | |
| DCAF1 | |
| A synthesized peptide derived from human DCAF1(Accession Q9Y4B6), corresponding to amino acid residues D30-K80. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction