Learn More
VPRBP Rabbit anti-Human, CoraLite 555, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech CL555-11612-100UL
Product Code. 17452613
  • € 450.00 / 100µL
Description
The VPRBP antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human VPRBP. This antibody recognizes human, mouse, rat antigen. The VPRBP antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™555.
Specifications
Specifications
VPRBP
Polyclonal
Rabbit
Primary
Human, Rat, Mouse
Immunofluorescence, Flow Cytometry (Intracellular Staining)
CoraLite 555
100 μL
9730