Learn More
Invitrogen™ VPS11 Recombinant Rabbit Monoclonal Antibody (24GB5260)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557318
This item is not returnable.
View return policy
Description
VPS11 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Vesicle mediated protein sorting plays an important role in segregation of intracellular molecules into distinct organelles. Genetic studies in yeast have identified more than 40 vacuolar protein sorting (VPS) genes involved in vesicle transport to vacuoles. This gene encodes the human homolog of yeast class C Vps11 protein. The mammalian class C Vps proteins are predominantly associated with late endosomes/lysosomes, and like their yeast counterparts, may mediate vesicle trafficking steps in the endosome/lysosome pathway. Alternative splicing results in multiple transcript variants.
Specifications
| VPS11 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| VPS11 | |
| 1200011A11Rik; END1; HLD12; hVPS11; PEP5; PP3476; RING finger protein 108; RNF108; vacuolar protein sorting 11 (yeast); vacuolar protein sorting 11 homolog; vacuolar protein sorting 11 homolog (S. cerevisiae); vacuolar protein sorting-associated protein 11 homolog; VPS11; VPS11 CORVET/HOPS core subunit; VPS11, CORVET/HOPS core subunit | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 315600, 55823, 71732 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB5260 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q91W86, Q9H270 | |
| VPS11 | |
| A synthesized peptide derived from human VPS11 (aa 10-44). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |