Learn More
Invitrogen™ VPS11 Recombinant Rabbit Monoclonal Antibody (24GB5260)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557317
 View more versions of this product
Product Code. 30275299
  • € 514.00 / 100µL
Description
VPS11 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Vesicle mediated protein sorting plays an important role in segregation of intracellular molecules into distinct organelles. Genetic studies in yeast have identified more than 40 vacuolar protein sorting (VPS) genes involved in vesicle transport to vacuoles. This gene encodes the human homolog of yeast class C Vps11 protein. The mammalian class C Vps proteins are predominantly associated with late endosomes/lysosomes, and like their yeast counterparts, may mediate vesicle trafficking steps in the endosome/lysosome pathway. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
VPS11
Recombinant Monoclonal
Unconjugated
VPS11
1200011A11Rik; END1; HLD12; hVPS11; PEP5; PP3476; RING finger protein 108; RNF108; vacuolar protein sorting 11 (yeast); vacuolar protein sorting 11 homolog; vacuolar protein sorting 11 homolog (S. cerevisiae); vacuolar protein sorting-associated protein 11 homolog; VPS11; VPS11 CORVET/HOPS core subunit; VPS11, CORVET/HOPS core subunit
Rabbit
Affinity chromatography
RUO
315600, 55823, 71732
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB5260
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q91W86, Q9H270
VPS11
A synthesized peptide derived from human VPS11 (aa 10-44).
100 μL
Primary
Human, Mouse, Rat