missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VPS13A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554489
This item is not returnable.
View return policy
Description
Immunogen sequence: RPPRFFNEDG VIRPYRLRDG TGNQMLQVME NGRFAKYKYF THVMINKTDM LMITRRGVLF VTKGTFGQLT CEWQYSFDEF TKEPFIVHGR RLRIEAKERV KSVF Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 34%.
Specifications
| VPS13A | |
| Polyclonal | |
| Unconjugated | |
| VPS13A | |
| CHAC; Chorea-acanthocytosis protein; Chorein; KIAA0986; vacuolar protein sorting 13 homolog A; vacuolar protein sorting 13A; vacuolar protein sorting-associated protein 13A; VPS13A | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23230 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96RL7 | |
| VPS13A | |
| Recombinant protein corresponding to Human VPS13A. Recombinant protein control fragment (Product #RP-93097). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction