missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VPS13B Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-82052CL1
This item is not returnable.
View return policy
Description
VPS13B Polyclonal antibody specifically detects VPS13B in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| VPS13B | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| cohen syndrome protein 1, DKFZp313I0811, KIAA0532, vacuolar protein sorting 13 homolog B (yeast) | |
| VPS13B antibody was raised against a 17 amino acid synthetic peptide near the amino terminus of human VPS13B. The immunogen is located within amino acids 90 - 140 of VPS13B. Amino Acid Squence: TNRSTAESTKSSIKPRR | |
| 0.1 mL | |
| Signal Transduction | |
| 157680 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction