missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VPS13B Antibody (OTI6F6), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74853B
This item is not returnable.
View return policy
Description
VPS13B Monoclonal specifically detects VPS13B in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| VPS13B | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| VPS13B | |
| Recombinant protein fragment corresponding to amino acids 91-346 of human VPS13B (NP_858047) produced in E.coli. | |
| 0.1 mL | |
| 157680.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI6F6 | |
| Western Blot | |
| cohen syndrome protein 1, DKFZp313I0811, KIAA0532, vacuolar protein sorting 13 homolog B (yeast) | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction