missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VPS18 Antibody (SR1024), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22137-50ul
This item is not returnable.
View return policy
Description
VPS18 Monoclonal antibody specifically detects VPS18 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence
Specifications
| VPS18 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Flow Cytometry, Immunofluorescence | |
| SR1024 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| hVPS18, KIAA1475PEP3, vacuolar protein sorting 18 homolog (S. cerevisiae), vacuolar protein sorting protein 18, vacuolar protein sorting-associated protein 18 homolog | |
| A synthesized peptide derived from human VPS18 (Uniprot #: Q9P253) | |
| 50 μg | |
| Autophagy, Cellular Markers | |
| 57617 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction