missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VPS26B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA598468
This item is not returnable.
View return policy
Description
VPS26B Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
In mammals, there are two paralogues of yeast Vps26, VPS26A and VPS26B. VPS26 is a component of the retromer complex composed of VPS26 (VPS26A or VPS26B), VPS29, VPS35, SNX1 and SNX2. VPS26A and VPS26B subunits define distinct retromer complexes. The retromer complex is important in recycling transmembrane receptors from endosomes to the trans-Golgi network (TGN).
Specifications
| VPS26B | |
| Polyclonal | |
| Unconjugated | |
| VPS26B | |
| 1810012I05Rik; 2310075A12Rik; AI848392; Pep8b; vacuolar protein sorting 26 homolog B (S. pombe); vacuolar protein sorting 26 homolog B (yeast); vacuolar protein sorting-associated protein 26B; Vesicle protein sorting 26B; VPS26 retromer complex component B; VPS26, retromer complex component B; Vps26b | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein G | |
| RUO | |
| 112936 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 3.19 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300, 0.02% sodium azide; pH 7.4 | |
| Q4G0F5 | |
| VPS26B | |
| Recombinant Human Vacuolar protein sorting-associated protein 26B protein (1-336AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction