missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VPS28 Antibody (OTI1A8), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-45412
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot, Immunohistochemistry, Immunofluorescence
VPS28 Monoclonal specifically detects VPS28 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence.
Specifications
| VPS28 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:200 - 1:2000, Immunohistochemistry 1:150, Immunofluorescence 1:100 | |
| ESCRT-I complex subunit VPS28, H-Vps28, MGC60323, vacuolar protein sorting 28 (yeast), vacuolar protein sorting 28 homolog (S. cerevisiae), vacuolar protein sorting-associated protein 28 homolog, yeast class E protein Vps28p homolog | |
| Mouse | |
| 25.2 kDa | |
| 0.1 mL | |
| Autophagy | |
| 51160 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| OTI1A8 | |
| Unconjugated | |
| PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
| VPS28 | |
| Full length human recombinant protein of human VPS28(NP_057292) produced in HEK293T cell. | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction