Learn More
Invitrogen™ VPS28 Recombinant Rabbit Monoclonal Antibody (24GB1755)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557844
 View more versions of this product
Product Code. 30276256
  • € 564.00 / 100µL
Description
VPS28 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot
This gene encodes a protein involved in endosomal sorting of cell surface receptors via a multivesicular body/late endosome pathway. The encoded protein is one of the three subunits of the ESCRT-I complex (endosomal complexes required for transport) involved in the sorting of ubiquitinated proteins. The two other subunits of ESCRT-I are vesicular protein sorting 23, also known as tumor susceptibility gene 101 (TSG101), and vesicular protein sorting 37. Two alternative transcripts encoding different isoforms have been described. Additional alternative transcripts may exist but the proteins encoded by these transcripts have not been verified experimentally.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
VPS28
Recombinant Monoclonal
Unconjugated
VPS28
1110014J03Rik; Caspase-activated DNase inhibitor that interacts with ASK1; CIIA; D730005C08Rik; ESCRT-I complex subunit VPS28; H-Vps28; MGC60323; vacuolar protein sorting 28 (yeast); vacuolar protein sorting 28 homolog; vacuolar protein sorting 28 homolog (S. cerevisiae); vacuolar protein sorting 28-like protein; vacuolar protein sorting-associated protein 28 homolog; VPS28; VPS28 subunit of ESCRT-I; VPS28, ESCRT-I subunit; yeast class E protein Vps28p homolog
Rabbit
Affinity chromatography
RUO
300052, 51160, 66914
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB1755
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
B5DEN9, Q9D1C8, Q9UK41
VPS28
A synthesized peptide derived from human beta VPS28 (aa 150-221).