missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VPS29 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-1387
This item is not returnable.
View return policy
Description
VPS29 Polyclonal specifically detects VPS29 in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| VPS29 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| DC15, DC7, DKFZp564F0223, EC 3.1.3.3, FLJ20492, hVPS29, PEP11, PEP11 homolog, retromer protein, vacuolar protein sorting 29 (yeast homolog), vacuolar protein sorting 29 (yeast), vacuolar protein sorting 29 homolog (S. cerevisiae), vacuolar protein sorting-associated protein 29, vacuolar sorting protein VPS29/PEP11, Vesicle protein sorting 29, x 007 protein | |
| Goat | |
| 21 kDa | |
| 0.1 mg | |
| Autophagy, Protein Phosphatase | |
| 51699 | |
| Human | |
| IgG |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.1 - 0.3 ug/ml, Peptide ELISA Detection limit 1:16000 | |
| Q9UBQ0 | |
| VPS29 | |
| Peptide with sequence C-DDVKVERIEYKKP corresponding to C-Terminus according to NP_057310.1, NP_476528.1. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This antibody is expected to recognize both human isoforms (represented by NP_057310.1; NP_476528.1). | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction