missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VPS29 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599024
This item is not returnable.
View return policy
Description
VPS29 Polyclonal Antibody for Western Blot, IHC (P), ELISA
This gene belongs to a group of vacuolar protein sorting genes that, when functionally impaired, disrupt the efficient delivery of vacuolar hydrolases. The protein encoded by this gene is a component of a large multimeric complex, termed the retromer complex, which is involved in retrograde transport of proteins from endosomes to the trans-Golgi network. This VPS protein may be involved in the formation of the inner shell of the retromer coat for retrograde vesicles leaving the prevacuolar compartment. Alternative splice variants encoding different isoforms, and usage of multiple polyadenylation sites have been found for this gene.
Specifications
| VPS29 | |
| Polyclonal | |
| Unconjugated | |
| VPS29 | |
| 2010015D08Rik; AW049835; DC15; DC7; DKFZp564F0223; EMBL:AAI67055.1, ECO:0000312; FLJ20492; hVPS29; MDS007; PEP11; PEP11 homolog; retromer protein; RGD:1308332}; UniProtKB:Q9UBQ0}; vacuolar protein sorting 29 (S. pombe); vacuolar protein sorting 29 homolog; vacuolar protein sorting 29 homolog (S. cerevisiae); vacuolar protein sorting-associated protein 29; vacuolar protein sorting-associated protein 29 {ECO:0000250; vacuolar sorting protein 29; vacuolar sorting protein VPS29/PEP11; vesicle protein sorting 29; vesicle protein sorting 29 {ECO:0000250; VPS29; vps29 {ECO:0000312; VPS29 retromer complex component; VPS29, retromer complex component; x 007 protein | |
| Rabbit | |
| Protein G | |
| RUO | |
| 51699 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Western Blot | |
| 5 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q9UBQ0 | |
| VPS29 | |
| Recombinant Human Vacuolar protein sorting-associated protein 29 protein (68-145AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction