missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VPS33B Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94816-0.02ml
This item is not returnable.
View return policy
Description
VPS33B Polyclonal antibody specifically detects VPS33B in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| VPS33B | |
| Polyclonal | |
| Western Blot 1:1000-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| FLJ14848, hVPS33B, vacuolar protein sorting 33 homolog B (yeast), vacuolar protein sorting 33B, vacuolar protein sorting 33B (yeast homolog), vacuolar protein sorting-associated protein 33B | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 348-617 of human VPS33B (NP_061138.3). CESIMKKKTKQDFQELIKTEHALLEGFNIRESTSYIEEHIDRQVSPIESLRLMCLLSITENGLIPKDYRSLKTQYLQSYGPEHLLTFSNLRRAGLLTEQAPGDTLTAVESKVSKLVTDKAAGKITDAFSSLAKRSNFRAISKKLNLIPRVDGEYDLKVPRDMAYVFGGAYVPLSCRIIEQVLERRSWQGLDEVVRLLNCSDFAFTDMTKEDKASSESLRLILVVFLGGCTFSEISALRFLGREKGYRFIFLTTAVTNSARLMEAMSEVKA | |
| 0.02 mL | |
| Signal Transduction | |
| 26276 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction