missing translation for 'onlineSavingsMsg'
Learn More

Vps34-PIK-III, MedChemExpress

Product Code. 16415358
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16415358

Brand: Medchem Express HY12794

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Vps34-PIK-III is a potent and selective inhibitor of VPS34 with an IC50 of 18 nM.

Vps34-PIK-III is a potent and selective inhibitor of VPS34 with an IC50 of 18 nM.

Specifications

Chemical Name or Material Vps34-PIK-III
CAS 1383716-40-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C17H17N7
Quantity 10 mg
Solubility Information DMSO : ≥ 31 mg/mL (97.07 mM)
SMILES NC1=NC=C(C2=CC=NC(NC3=CC=NC=C3)=N2)C(CC4CC4)=N1
Molecular Weight (g/mol) 319.36
Formula Weight 319.36
Percent Purity 99.02%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.