missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VPS34 Recombinant Rabbit Monoclonal Antibody (HL2305)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA550485
This item is not returnable.
View return policy
Description
VPS34 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
PIK3C3 (hVPS34) is the sole class III member of the PI3 Kinase family. PIK3C3 phosphorylates phosphatidylinositol (PI) to produce phosphatidylinositol 3-phosphate [PI(3)P], which recruits downstream effectors containing FYVE and Phox domains to membranes. PIK3C3 is involved in coupling nutrient signaling to mTOR and also plays a role in autophagy and intracellular trafficking. Involved in regulation of degradative endocytic trafficking and required for the abcission step in cytokinesis, probably in the context of PI3KC3-C2. Involved in the transport of lysosomal enzyme precursors to lysosomes. Required for transport from early to late endosomes.
Specifications
| VPS34 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| O88763, Q6PF93, Q8NEB9 | |
| Pik3c3 | |
| Recombinant fragmemt of human VPS34. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Canine | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| HL2305 | |
| Unconjugated | |
| Pik3c3 | |
| 5330434F23Rik; catalytic phosphatidylinositol 3-kinase 3; catalytic subunit type 3 Antibody; class 3 phosphoinositide-3-kinase; Class III PI 3-kinase; hVps34; hVps34 Antibody; OTTMUSP00000023341; OTTMUSP00000023350; phosphatidylinositol 3-kinase; Phosphatidylinositol 3-kinase catalytic subunit type 3; phosphatidylinositol 3-kinase p100 subunit; phosphatidylinositol 3-kinase, catalytic subunit type 3; Phosphoinositide-3-kinase class 3; phosphoinositide-3-kinase, class 3; PI3K type 3; PI-3K Vps34p; pi3kc3; PI3-kinase type 3; Pik3c3; PIK3C3 Antibody; PtdIns-3-kinase type 3; VPS34 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 225326, 490477, 5289, 65052 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction