missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VPS35 Antibody (S03-6E1), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19926-50ul
This item is not returnable.
View return policy
Description
VPS35 Monoclonal antibody specifically detects VPS35 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| VPS35 | |
| Monoclonal | |
| 0.3 mg/mL | |
| Western Blot, Immunohistochemistry 1:20, Immunohistochemistry-Paraffin | |
| DKFZp434P1672, FLJ10752, FLJ13588, FLJ20388, hVPS35, Maternal-embryonic 3, MEM3DKFZp434E1211, vacuolar protein sorting 35 (yeast homolog), vacuolar protein sorting 35 (yeast), vacuolar protein sorting 35 homolog (S. cerevisiae), vacuolar protein sorting-associated protein 35, Vesicle protein sorting 35 | |
| A synthetic peptide of human VPS35 (Uniprot # Q96QK1) | |
| 50 μg | |
| Autophagy, Cellular Markers | |
| 55737 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| S03-6E1 | |
| Unconjugated | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction