missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VPS37B Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-06284CL1
This item is not returnable.
View return policy
Description
VPS37B Polyclonal antibody specifically detects VPS37B in Human samples. It is validated for Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| VPS37B | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
| CoraFluor 1 | |
| ESCRT-I complex subunit VPS37B, FLJ12750, hVps37B, vacuolar protein sorting 37 homolog B (S. cerevisiae), vacuolar protein sorting 37B, vacuolar protein sorting 37B (yeast), vacuolar protein sorting-associated protein 37B | |
| Produced in rabbits immunized with E. coli-derived Human VPS37B fragment. | |
| 0.1 mL | |
| Signal Transduction | |
| 79720 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?