missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VPS37B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5118354
This item is not returnable.
View return policy
Description
VPS37B Polyclonal Antibody for Western Blot, IHC (P), IP
Component of the ESCRT-I complex, a regulator of vesicular trafficking process. Required for the sorting of endocytic ubiquitinated cargos into multivesicular bodies. May be involved in cell growth and differentiation.
Specifications
| VPS37B | |
| Polyclonal | |
| PBS with 0.03% ProClin 300; pH 7 | |
| Q9H9H4 | |
| VPS37B | |
| E. coli-derived Human VPS37B fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| VPS37B | |
| 2300007F24Rik; AI415429; BC026744; ESCRT-I complex subunit VPS37B; hVps37B; RGD1304576; vacuolar protein sorting 37 homolog B; vacuolar protein sorting 37 homolog B (S. cerevisiae); vacuolar protein sorting 37B; vacuolar protein sorting 37B (yeast); vacuolar protein sorting-associated protein 37B; Vps37b; VPS37B, ESCRT-I subunit | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 79720 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction