missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ VPS39 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16557031
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16557031 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16557031 Supplier Invitrogen™ Supplier No. PA588969

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: RFHNCLIQLY CEKVQGLMKE YLLSFPAGKT PVPAGEEEGE LGEYRQKLLM FLEISSYYDP GRLICDFPFD GLLEERALLL GRMGKHEQAL FIYVHILKDT RMAEEYCHKH YDRNKDGNKD VYLSLLRMYL SPPSIHCLGP IKLELLEPKA NLQAALQVLE LHHSKLDTTK ALNLLPANTQ INDIRIFLEK VLEENAQKKR FNQVLKNLLH AEFLRVQEER ILHQQVKCII TEEKVCMVCK KKIGNSAFAR YPNGVVVHYF CSKEVNPADT; Positive Samples: U-87MG, 293T, A-549; Cellular Location: Cytoplasm, Late endosome, Late endosome membrane, Lysosome, Lysosome membrane, Peripheral membrane protein.

VPS39 is the mammalian homolog of the yeast vacuolar protein VAM6, that in mammals, promotes lysosome clustering and fusion in vivo. VPS39 is also thought to modulate the TGF-beta response by coupling the TGF-beta receptor complex to the Smad pathway. Similar to the related protein TRAP1, the VPS39 protein is essential for early embryonic development, with mice lacking VPS39 dying at or before E6. 5.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VPS39
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.01 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene VPS39
Gene Accession No. Q8R5L3, Q96JC1
Gene Alias A230065P22Rik; AW743070; hVam6p; KIAA0770; mVam6; OTTMUSP00000023341; OTTMUSP00000023350; PI3K type 3; Pldn; TLP; TRAP1-like protein; vacuolar protein sorting 39 (yeast); vacuolar protein sorting 39 homolog; vacuolar protein sorting 39 homolog (S. cerevisiae); Vam6; Vam6/Vps39-like protein; Vam6P; Vps39; VPS39 HOPS complex subunit; VPS39, HOPS complex subunit
Gene Symbols VPS39
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 606-875 of human VPS39 (NP_0561042).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23339, 269338, 362199
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.