missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VPS45 Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98574MFV500
This item is not returnable.
View return policy
Description
VPS45 Polyclonal antibody specifically detects VPS45 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| VPS45 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| H1, H1VPS45, hlVps45, h-vps45, leucocyte vacuolar protein sorting 45, vacuolar protein sorting 45 homolog (S. cerevisiae), vacuolar protein sorting 45A, vacuolar protein sorting 45A (yeast homolog), vacuolar protein sorting 45A (yeast), vacuolar protein sorting-associated protein 45, VPS45A, VPS45AVPS54A, VPS45B, VSP45, VSP45A | |
| Produced in rabbits immunized with E. coli-derived Human VPS45 fragment. | |
| 0.1 mL | |
| Cellular Markers, Golgi Apparatus Markers | |
| 11311 | |
| Store at 4C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction