missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VPS50 Monoclonal Antibody (OTI7A8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies CF812028
This item is not returnable.
View return policy
Description
VPS50 Monoclonal antibody specifically detects VPS50 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| VPS50 | |
| Monoclonal | |
| See Label | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| O95837, P30677 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 14675, 309242, 9630 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI7A8 | |
| Unconjugated | |
| Gna14 | |
| 1700034M03Rik; 8430415E05Rik; Bcl2-like protein blm; BLM; C87205; CCDC132; coiled-coil domain containing 132; coiled-coil domain-containing protein 132; DKFZp313I2429; EARP/GARPII complex subunit VPS50; FLJ20097; KIAA1861; mKIAA1861; RGD1304751; syndetin; Vps50; VPS50 EARP/GARPII complex subunit; VPS50, EARP/GARPII complex subunit; VPS54L | |
| Human recombinant protein fragment corresponding to amino acids 31-348 of human GNA14 (NP_004288) produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Lyophilized |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction