missing translation for 'onlineSavingsMsg'
Learn More

VPS51 Antibody, CoraFluor™ 1, Novus Biologicals™

Product Code. 30151638
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30151638 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30151638 Supplier Novus Biologicals (Bio-Techne) Supplier No. NBP282053CL1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

VPS51 Polyclonal antibody specifically detects VPS51 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen VPS51
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate CoraFluor 1
Formulation PBS
Gene Alias ANG2, ANG3, C11orf2, C11orf3, chromosome 11 open reading frame 2, FFR, protein fat-free homolog, vacuolar protein sorting 51 homolog (S. cerevisiae)
Host Species Rabbit
Immunogen VPS51 antibody was raised against an 18 amino acid synthetic peptide near the carboxy terminus of human VPS51. The immunogen is located within amino acids 620 - 670 of VPS51. Amino Acid Squence: LYEEGVRKAQSSDSSKRT
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 738
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.