Learn More
VPS51 Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-82053MFV450
This item is not returnable.
View return policy
Description
VPS51 Polyclonal antibody specifically detects VPS51 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
Specifications
| VPS51 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Store at 4C in the dark. |
| Western Blot | |
| mFluor Violet 450 SE | |
| ANG2, ANG3, C11orf2, C11orf3, chromosome 11 open reading frame 2, FFR, protein fat-free homolog, vacuolar protein sorting 51 homolog (S. cerevisiae) |