missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VPS52 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97979-100ul
This item is not returnable.
View return policy
Description
VPS52 Polyclonal antibody specifically detects VPS52 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| VPS52 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| ARE1, dJ1033B10.5, DKFZp547I194, SAC2, SAC2 suppressor of actin mutations 2-like (yeast), SAC2 suppressor of actin mutations 2-like protein, SACM2LRP5-1033B10, vacuolar protein sorting 52 (yeast), vacuolar protein sorting 52 homolog (S. cerevisiae), vacuolar protein sorting-associated protein 52 homolog | |
| Produced in rabbits immunized with E. coli-derived Human VPS52 fragment. | |
| 100 μg | |
| Cellular Markers | |
| 6293 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction