missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VPS52 Polyclonal Antibody

VPS52 Polyclonal Antibody
Brand: Thermo Scientific PA560071
This item is not returnable.
View return policy
Description
VPS52 Polyclonal Antibody for IHC
Specifications
| VPS52 | |
| Polyclonal | |
| Unconjugated | |
| VPS52 | |
| Are1; D130068D18; D430041K17Rik; dJ1033B10.5; DKFZp547I194; fb94c11; LOW QUALITY PROTEIN: vacuolar protein sorting-associated protein 52 homolog; RP5-1033B10; SAC2; SAC2 suppressor of actin mutations 2-like protein; SACM2L; tclw5; vacuolar protein sorting 52 (yeast); vacuolar protein sorting 52 homolog; vacuolar protein sorting 52 homolog (S. cerevisiae); vacuolar protein sorting-associated protein 52 homolog; vacuolar protein sorting-associated protein 52-like protein; Vacuolar protein sorting-associated protein 52-like protein-like protein; Vps52; VPS52 GARP complex subunit; VPS52 subunit of GARP complex; VPS52, GARP complex subunit; wu:fb94c11 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6293 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.8 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8N1B4 | |
| VPS52 | |
| Recombinant protein corresponding to Human VPS52. Recombinant protein control fragment (Product #RP-99254). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction