Learn More
VPS53 Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP1-76901G
Product Code. 18494934
  • € 582.00 / 0.10mL
Description
VPS53 Polyclonal antibody specifically detects VPS53 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
VPS53
Polyclonal
50mM Sodium Borate
Rabbit
Peptide affinity purified
RUO
55275
Store at 4°C in the dark.
IgG
Western Blot, ELISA, Immunofluorescence
DyLight 488
FLJ10979, FLJ41112, FLJ61757, HCCS1, hVps53L, MGC39512, pp13624, vacuolar protein sorting 53 (yeast), vacuolar protein sorting 53 homolog (S. cerevisiae), vacuolar protein sorting-associated protein 53 homolog
Antibody was raised against a 18 amino acid synthetic peptide from near the carboxy terminus of human VPS53. The immunogen is located within the last 50 amino acids of VPS53. Amino Acid Squence: LTAPTPEQESSRIRKLEK
0.1 mL
Primary
Human, Mouse, Rat