missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ VPS53 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15702591 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15702591 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15702591 Supplier Invitrogen™ Supplier No. PA554656

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: LRDACLVANI LDPRIKQEII KKFIKQHLSE YLVLFQENQD VAWLDKIDRR YAWIKRQLVD YEEKYGRMFP REWCMAERIA VEFCHVTRAE LAKIMRTRAK EIEVKLL Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 94%.

The sorting of acid hydrolases to lysosomes rely on mannose 6-phosphate receptors that cycle between the trans-Golgi network (TGN) and endosomes. The maintenance of this cycle requires the function of the mammalian Golgi-associated retrograde protein (GARP) complex which is composed of three subunits: VPS52, VPS53, and VPS54. Depletion of any of these three proteins, such as by RNAi, impairs the retrograde transport of multiple TGN proteins. VPS53 was identified as an HIV dependency factor (HDF) and plays a role in viral entry to the cell, suggesting that VPS53 may be an important drug target in HIV treatment. At least five isoforms of VPS53 are known to exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VPS53
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Vps53
Gene Accession No. Q5VIR6
Gene Alias 2010002A08Rik; 2310040I21Rik; 3100002B05Rik; Are1; Hccs1; hVps53L; PCH2E; pp13624; RGD1311391; SAC2 suppressor of actin mutations 2-like protein; Sacm2l; vacuolar protein sorting 52 homolog (S. cerevisiae); vacuolar protein sorting 53 (yeast); vacuolar protein sorting 53 homolog (S. cerevisiae); vacuolar protein sorting-associated protein 52 homolog; Vacuolar protein sorting-associated protein 53 homolog; Vps52; Vps53; VPS53 GARP complex subunit; VPS53, GARP complex subunit
Gene Symbols Vps53
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human VPS53. Recombinant protein control fragment (Product #RP-93279).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55275
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.