missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VPS72 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | VPS72 |
|---|---|
| Concentration | 0.25 mg/mL |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16854194
|
Proteintech
15143-1-AP-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16844194
|
Proteintech
15143-1-AP-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Vacuolar protein sorting-associated protein 72 homolog (VPS72), also known as VPS72 or Transcription factor-like 1, is a 364 amino acid subunit of the TRRAP/TIP60 HAT complex. VPS72 has also been identified as a subunit of a novel complex containing SNF2-related helicase SRCAP (SWI2/SNF2-related CBP activator protein). This SRCAP-containing complex is very similar to the S. cerevisiae SWR1 chromatin remodeling complex. The involvement of VPS72 in these complexes has suggested that VPS72 plays multiple roles in chromatin modification and remodeling in cells. VPS72 localizes to the nucleus and is phosphorylated upon DNA damage, most likely by ATM or ATR.Specifications
| VPS72 | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Mouse, Rat, Human | |
| Q15906, Q62481 | |
| 21427, 310661, 6944 | |
| VPS72 Fusion Protein Ag7424 | |
| Primary | |
| -20°C | |
| VPS72 |
| 0.25 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| CFL1, Protein YL 1, Swc2, TCFL1, Transcription factor like 1, VPS72, YL 1, YL1 | |
| VPS72 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title