missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VPS9D1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552472
This item is not returnable.
View return policy
Description
Immunogen sequence: EAYTEYLRSI HYISQVLLEE VETTKEAGET VPPDTSKMLK LAQQCLERAQ STAAKLGKTR LKPTMPAAAP IPQPAGRHRR VYSDEGGKLS PFLPPEIFQK LQGAESQSCK K Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 79%.
Specifications
| VPS9D1 | |
| Polyclonal | |
| Unconjugated | |
| VPS9D1 | |
| 1300018I17Rik; 2410004N05Rik; 5-day ovary-specific transcript 1 protein; 5DOS1; ATPBL; ATP-BL; C16orf7; Protein ATP-BL; RGD1565149; VPS9 domain containing 1; VPS9 domain-containing protein 1; VPS9D1; VSP9 domain containing 1; Vsp9d1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9605 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y2B5 | |
| VPS9D1 | |
| Recombinant protein corresponding to Human VPS9D1. Recombinant protein control fragment (Product #RP-89297). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction