missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VRK1 Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99819MFV610
This item is not returnable.
View return policy
Description
VRK1 Polyclonal antibody specifically detects VRK1 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation
Specifications
| ELISA | |
| mFluor Violet 610 SE | |
| Produced in rabbits immunized with purified, recombinant Human VRK1 (Uniprot#: Q99986; Met1-Lys396) | |
| 7443 |
| Polyclonal | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction