missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VRK1 Antibody (OTI2C9), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-46351
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot
VRK1 Monoclonal specifically detects VRK1 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| VRK1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:2000 | |
| EC 2.7.11.1, MGC117401, MGC138280, MGC142070, PCH1, serine/threonine-protein kinase VRK1, vaccinia related kinase 1, vaccinia virus B1R-related kinase 1, Vaccinia-related kinase 1 | |
| Mouse | |
| 45.3 kDa | |
| 0.1 mL | |
| Core ESC Like Genes, Protein Kinase, Stem Cell Markers | |
| 7443 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot | |
| OTI2C9 | |
| Unconjugated | |
| PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
| VRK1 | |
| Human recombinant protein fragment corresponding to amino acids 1-230 of human VRK1(NP_003375) produced in E.coli. | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction